Proteintech
Manufacturer Code:139381AP150UL
Catalog # 50-172-9105
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Calponin Fusion Protein Ag4989 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acidic calponin h3; Basic calponin; Basic calponin Calponin Calponin 1 Calponin H1 smooth muscle CNN1 Sm Calp SMCC; calponin 1, basic, smooth muscle; calponin 1, smooth muscle; calponin 2; calponin 3, acidic; Calponin H1, smooth muscle; Calponin H2, smooth muscle; Calponin, acidic isoform; Calponin, non-muscle isoform; Calponin-1; Calponin-2; Calponin-3; calponins, basic; dJ639P13.2.2 (acidic calponin 3); epididymis secretory protein Li 14; h2-calponin; Neutral calponin
Gene Aliases: 1600014M03Rik; AA408047; AI324678; C85854; Calpo2; Calpo3; CN; CNN1; CNN2; CNN3; CnnI; HEL-S-14; Sm-Calp; SMCC
UniProt ID: (Human) P51911, (Human) Q99439, (Human) Q15417, (Mouse) Q08091, (Rat) Q08290, (Mouse) Q08093, (Mouse) Q9DAW9, (Rat) P37397
Entrez Gene ID: (Human) 1264, (Human) 1265, (Human) 1266, (Mouse) 12797, (Rat) 65204, (Mouse) 12798, (Rat) 690976, (Mouse) 71994, (Rat) 54321
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.