Novus Biologicals
Manufacturer Code:NBP270421JF646
Catalog # N270421J646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B5 |
Immunogen | Full length recombinant protein of human Calponin 2 (NP_004359) produced in HEK293T cells. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: calponin 2 Calponin H2 smooth muscle calponin-2 Neutral calponin; Calponin H2, smooth muscle; Calponin-2; Neutral calponin
Gene Aliases: CNN2
UniProt ID: (Human) Q99439
Entrez Gene ID: (Human) 1265
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.