Invitrogen
Manufacturer Code:PA543135
Catalog # PIPA543135
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human CNN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Basic calponin; Calponin 1 basic smooth muscle SMCC Sm-Calp; calponin 1, basic, smooth muscle; calponin 1, smooth muscle; Calponin H1, smooth muscle; Calponin-1; calponins, basic; CNN1; epididymis secretory protein Li 14; h1-calponin; smooth muscle calponin h1
Gene Aliases: CN; CNN1; CnnI; HEL-S-14; Sm-Calp; SMCC
UniProt ID: (Human) P51911, (Sheep) Q7YRL2, (Bovine) Q2HJ38, (Pig) Q08092, (Mouse) Q08091, (Rat) Q08290
Entrez Gene ID: (Human) 1264, (Sheep) 443517, (Bovine) 534583, (Horse) 100054768, (Pig) 396911, (Dog) 484937, (Mouse) 12797, (Rat) 65204
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.