Novus Biologicals
Manufacturer Code:NBP190329
Catalog # NBP190329
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:THYSNIEANESEEVRQFRRLFAQLAGDDMEVSATELMNILNKVVTRHPDLKTDGFGIDTCRSMVAVMDSDTTGKLGFEEFKYLWNNIKRWQAIYKQFDTDRSGTICSSELPG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-activated neutral proteinase small subunit; Calcium-activated neutral proteinase small subunit Calcium-dependent protease small subunit Calcium-dependent protease small subunit 1 calcium-dependent protease small subunit calpain 4 small subunit (30K) Calpain regulatory subunit calpain small subunit 1 calpain small polypeptide calpain small subunit 1 CANP CANPS CAPN4CALPAIN430K CAPNS CDPSCANP small subunit CSS1; Calcium-dependent protease small subunit; Calcium-dependent protease small subunit 1; calcium-dependent protease, small subunit; calpain 4, small subunit (30K); Calpain regulatory subunit; Calpain small subunit 1; calpain, small polypeptide; calpain, small subunit 1; CANP small subunit; CDPS; CSS1
Gene Aliases: CALPAIN4; CANP; CANPS; CAPN4; CAPNS; CAPNS1; CDPS; CSS1
UniProt ID: (Human) P04632
Entrez Gene ID: (Human) 826
Molecular Function: annexin calcium-binding protein calmodulin cysteine protease hydrolase intracellular calcium-sensing protein protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.