Novus Biologicals
Manufacturer Code:NBP214896
Catalog # NBP214896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human Calpain 9. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calpain 9 calpain 9 (nCL-4) calpain-9 Digestive tract-specific calpain EC 3.4.22 EC 3.4.22.54 GC36 NCL4 nCL-4EC 3.4.22.- New calpain 4 novel calpain large subunit-4 Protein CG36; Calpain-9; Digestive tract-specific calpain; nCL-4; New calpain 4; novel calpain large subunit-4; Protein CG36
Gene Aliases: CAPN9; GC36; nCL-4; NCL4
UniProt ID: (Human) O14815
Entrez Gene ID: (Human) 10753
Molecular Function: annexin calcium-binding protein calmodulin cysteine protease hydrolase intracellular calcium-sensing protein protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.