Novus Biologicals
Manufacturer Code:NBP214436
Catalog # NBP214436
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: AVFYYKEAAQALIYAEMAGSSLENIQEKITEYLERVQALHSAVQSKSADP LKSKHQLDLERAHFLVTQAFDEDEKENVEDAIELYTEAVDLCLKTSYETA DKVLQNK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calpain 7 calpain like protease CALPAIN7 EC 3.4.22 EC 3.4.22.- FLJ36423 homolog of Aspergillus Nidulans PALB PalB homolog PalBH PALBHcalpain-7; calpain like protease; Calpain-7; homolog of Aspergillus Nidulans PALB; PalB homolog; PalBH
Gene Aliases: CALPAIN7; CAPN7; PALBH
UniProt ID: (Human) Q9Y6W3
Entrez Gene ID: (Human) 23473
Molecular Function: annexin calcium-binding protein calmodulin cysteine protease hydrolase intracellular calcium-sensing protein protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.