Proteintech
Manufacturer Code:269851AP150UL
Catalog # 50-173-5526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Calpain 7 Fusion Protein Ag25574 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calpain 7 CALPAIN7 CAPN7 PalB homolog PALBH; calpain like protease; Calpain-7; homolog of Aspergillus Nidulans PALB; PalB homolog; PalBH
Gene Aliases: AU022319; CALPAIN7; CAPN7; PALBH
UniProt ID: (Human) Q9Y6W3, (Mouse) Q9R1S8
Entrez Gene ID: (Human) 23473, (Mouse) 12339
Molecular Function: annexin calcium-binding protein calmodulin cysteine protease hydrolase intracellular calcium-sensing protein protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.