Novus Biologicals
Manufacturer Code:NBP292674100UL
Catalog # NB027217
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 361-640 of human CAPN5 (NP_004046.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calpain 5 Calpain htra-3 calpain-5 EC 3.4.22 EC 3.4.22.54 FLJ46245 HTRA3NCL3 nCL-3EC 3.4.22.- New calpain 3; Calpain htra-3; Calpain-5; nCL-3; New calpain 3; testis tissue sperm-binding protein Li 91mP
Gene Aliases: ADNIV; CAPN5; HTRA3; nCL-3; NCL3; VRNI
UniProt ID: (Human) O15484
Entrez Gene ID: (Human) 726
Molecular Function: annexin calcium-binding protein calmodulin cysteine protease hydrolase intracellular calcium-sensing protein protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.