Novus Biologicals
Manufacturer Code:NBP287117
Catalog # NB034739
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Calpain 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-activated neutral proteinase 3; calpain 3 (p94) Calpain L3 Calpain p94 calpain large polypeptide L3 calpain-3 CANP 3 CANP3MGC4403 CANPL3 EC 3.4.22 EC 3.4.22.54 LGMD2 LGMD2A MGC10767 MGC11121 MGC14344 Muscle-specific calcium-activated neutral protease 3 muscle-specific calcium-activated neutral protease 3 large subunit NCL1 nCL-1large [catalytic] subunit New calpain 1; calpain 3, (p94); Calpain L3; Calpain p94; calpain p94, large [catalytic] subunit; calpain, large polypeptide L3; Calpain-3; CANP 3; Muscle-specific calcium-activated neutral protease 3; muscle-specific calcium-activated neutral protease 3 large subunit; nCL-1; New calpain 1
Gene Aliases: CANP3; CANPL3; CAPN3; LGMD2; LGMD2A; nCL-1; NCL1; p94
UniProt ID: (Human) P20807
Entrez Gene ID: (Human) 825
Molecular Function:
annexin
calcium-binding protein
calmodulin
cysteine protease
hydrolase
intracellular calcium-sensing protein
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.