Proteintech
Manufacturer Code:104151AP150UL
Catalog # 50-172-6290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Calpain 3 Fusion Protein Ag0671 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-activated neutral proteinase 3; Calpain 3 calpain 3 (p94) Calpain L3 Calpain p94 CANP 3 CANP3 CANPL3 CAPN3 LGMD2 LGMD2A nCL 1 NCL1 New calpain 1 p94; calpain 3, (p94); Calpain L3; Calpain p94; calpain p94, large [catalytic] subunit; calpain, large polypeptide L3; Calpain-3; CANP 3; Muscle-specific calcium-activated neutral protease 3; muscle-specific calcium-activated neutral protease 3 large subunit; nCL-1; New calpain 1; skeletal muscle specific calpain p94
Gene Aliases: AI323605; CANP3; CANPL3; Capa-3; Capa3; CAPN3; LGMD2; LGMD2A; Lp82; Lp84; Lp85; nCL-1; NCL1; p94
UniProt ID: (Human) P20807, (Mouse) Q64691, (Rat) P16259
Entrez Gene ID: (Human) 825, (Mouse) 12335, (Rat) 29155
Molecular Function:
annexin
calcium-binding protein
calmodulin
cysteine protease
hydrolase
intracellular calcium-sensing protein
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.