Novus Biologicals
Manufacturer Code:NBP215675
Catalog # NBP215675
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Calpain 2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-activated neutral proteinase 2; Calcium-activated neutral proteinase 2 calpain 2 (m/II) large subunit calpain 2 large [catalytic] subunit calpain 2 large subunit Calpain large polypeptide L2 Calpain M-type calpain large polypeptide L2 calpain-2 catalytic subunit Calpain-2 large subunit CANP 2 CANPL2CANP2 CANPml EC 3.4.22 EC 3.4.22.53 FLJ39928 M-calpain mCANP Millimolar-calpain; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; Calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; Calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; M-calpain; Millimolar-calpain
Gene Aliases: CANP2; CANPL2; CANPml; CAPN2; mCANP
UniProt ID: (Human) P17655
Entrez Gene ID: (Human) 824
Molecular Function: annexin calcium-binding protein calmodulin cysteine protease hydrolase intracellular calcium-sensing protein protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.