Proteintech
Manufacturer Code:105381AP150UL
Catalog # 50-172-6387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Calpain 1 Fusion Protein Ag0810 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-activated neutral proteinase 1; Calpain 1 Calpain 1 catalytic subunit Calpain 1 large subunit Calpain mu type CANP CANP 1 CANP1 CANPL1 CAPN1 Micromolar calpain muCANP muCL; calpain 1, (mu/I) large subunit; Calpain mu-type; calpain, large polypeptide L1; Calpain-1 catalytic subunit; Calpain-1 large subunit; CANP 1; Cell proliferation-inducing gene 30 protein; cell proliferation-inducing protein 30; Micromolar-calpain; mu-calpain; muCANP
Gene Aliases: CANP; CANP1; CANPL1; Capa-1; Capa1; CAPN1; Cls1; mu-calpin; muCANP; muCL; PIG30; SPG76
UniProt ID: (Human) P07384, (Mouse) O35350, (Rat) P97571
Entrez Gene ID: (Human) 823, (Mouse) 12333, (Rat) 29153
Molecular Function:
annexin
calcium-binding protein
calmodulin
cysteine protease
hydrolase
intracellular calcium-sensing protein
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.