Invitrogen
Manufacturer Code:MA531501
Catalog # PIMA531501
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT1563 |
Immunogen | Recombit protein encompassing a sequence within the center region of human Calnexin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Calnexin; IP90; Major histocompatibility complex class I antigen-binding protein p88; p90; P90 Antibody CNX Antibody IP90 Antibody CANX Antibody calnexin Antibody
Gene Aliases: 1110069N15Rik; AI988026; CANX; CNX; D11Ertd153e; IP90; P90
UniProt ID: (Human) P27824, (Rat) P35565, (Mouse) P35564
Entrez Gene ID: (Human) 821, (Rat) 29144, (Mouse) 12330
Molecular Function: calcium-binding protein chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.