Novus Biologicals
Manufacturer Code:NBP236570SS
Catalog # NBP236570SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C2.2D11 |
Immunogen | Partial recombinant human Calnexin protein (between amino acids 1-300). [UniProt P27824] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Calnexin; calnexin CNX IP90FLJ26570 Major histocompatibility complex class I antigen-binding protein p88 P90; IP90; Major histocompatibility complex class I antigen-binding protein p88; p90
Gene Aliases: CANX; CNX; IP90; P90
UniProt ID: (Human) P27824
Entrez Gene ID: (Human) 821
Molecular Function: calcium-binding protein chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.