Invitrogen
Manufacturer Code:PA577839
Catalog # PIPA577839
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Dog Calnexin C-terminal synthetic peptide conjugated to KLH. Identical to human mouse and rat calnexin sequences over these residues. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calmegin; Calnexin; Calnexin CALX&lowbarHUMAN CANX CNX FLJ26570 Histocompatibility complex class I antigen binding protein p88 IP90 Major histocompatibility complex class I antigen-binding protein p88 P90; IP90; Major histocompatibility complex class I antigen-binding protein p88; p90; pp90
Gene Aliases: 1110069N15Rik; AI988026; CANX; canx-a; canx-b; clgn; clnx; CNX; D11Ertd153e; IP90; P90; PP90
UniProt ID: (Dog) P24643, (Human) P27824, (Rat) P35565, (Mouse) P35564
Entrez Gene ID: (Chicken) 416288, (Dog) 403908, (Bovine) 407129, (Rabbit) 100341645, (Sheep) 101122273, (Pig) 100519184, (Human) 821, (Rat) 29144, (Guinea pig) 100726237, (Mouse) 12330, (Western clawed frog) 448161
Molecular Function: calcium-binding protein chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.