Invitrogen
Manufacturer Code:PA534665
Catalog # PIPA534665
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 243 and 418 of Calnexin (Uniprot IDNo.P27824) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calnexin; CNX IP90 P90 major histocompatibility complex class I antigen-binding protein p88; IP90; Major histocompatibility complex class I antigen-binding protein p88; p90
Gene Aliases: 1110069N15Rik; AI988026; CANX; CNX; D11Ertd153e; IP90; P90
UniProt ID: (Human) P27824, (Mouse) P35564
Entrez Gene ID: (Human) 821, (Mouse) 12330
Molecular Function:
calcium-binding protein
chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.