Novus Biologicals
Manufacturer Code:NBP267413
Catalog # NBP267413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SJ16-09 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: CALM CALM2 CALM3 CALML2 calmodulin calmodulin 1 (phosphorylase kinase delta) CaM CAM1 CAM2 CAM3 CAMB CAMC CAMI CAMIII DD132 PHKDCAM phosphorylase kinase delta subunit; calmodulin 1 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1
Gene Aliases: CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CAMB; CAMC; CAMI; CAMIII; CPVT4; DD132; LQT14; PHKD
UniProt ID: (Human) P0DP23
Entrez Gene ID: (Human) 801
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.