Invitrogen
Manufacturer Code:PA578374
Catalog # PIPA578374
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Full length Human Calmodulin recombit protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CALM1 CALML2 CAMI CPVT4 DD132 PHKD caM calmodulin 1 (phosphorylase kinase delta); calmodulin; Calmodulin 1 (phosphorylase kinase, delta); calmodulin 2 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; caM; LP7057 protein; phosphorylase kinase delta; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1; prepro-calmodulin 2
Gene Aliases: 1500001E21Rik; AI256814; AI327027; AI461935; AL024000; AL024017; CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CAMB; CAMC; CAMI; CAMII; CAMIII; CPVT4; DD132; LQT14; LQT15; PHKD; PHKD2
UniProt ID: (Human) P0DP23, (Rat) P0DP29, (Mouse) P0DP26
Entrez Gene ID: (Human) 801, (Human) 805, (Rat) 24242, (Mouse) 12313, (Rat) 50663, (Mouse) 12314
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.