Invitrogen
Manufacturer Code:PA589886
Catalog # PIPA589886
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CALM3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calmodulin; Calmodulin III; Calmodulin-1; Calmodulin-2; Calmodulin-3; caM
Gene Aliases: Calm; Calm1; Calm2; Calm3; Cam; Cam1; Cam2; Cam3; CaMA; Camb; CamC; CaMI; CaMII; CaMIII; R75142; RNCAMIII
UniProt ID: (Rat) P0DP29, (Mouse) P0DP26
Entrez Gene ID: (Rat) 24244, (Mouse) 12315
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.