Invitrogen
Manufacturer Code:PA529980
Catalog # PIPA529980
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 99 of Human Calmodulin 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CALM1 CALM2 PHKD PHKD3 caM calmodulin; calmodulin; Calmodulin-1; Calmodulin-2; Calmodulin-3; caM
Gene Aliases: Calm; Calm1; Calm2; Calm3; Cam; Cam1; Cam2; Cam3; CaMA; CamC; R75142
UniProt ID: (Mouse) P0DP26
Entrez Gene ID: (Mouse) 12315
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.