Novus Biologicals
Manufacturer Code:NBP19847420UL
Catalog # NBP19847420
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is Calmodulin 3 - N-terminal region. Peptide sequence LQDMINEVDADGNGTIDFPEFLTMMARKMKDTDSEEEIREAFRVFDKDGN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CALM CALM1 CALM2 CALML2 calmodulin calmodulin 3 (phosphorylase kinase delta) CAM CAM1 CAM2 CAM3 CAMB CAMC CAMIII PHKD PHKD3; calmodulin 3 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; caM; epididymis secretory protein Li 72; phosphorylase kinase subunit delta; prepro-calmodulin 3
Gene Aliases: CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CAMB; CAMC; CAMIII; HEL-S-72; PHKD; PHKD3
UniProt ID: (Human) P0DP23
Entrez Gene ID: (Human) 808
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.