Novus Biologicals
Manufacturer Code:NBP299992G
Catalog # NB090587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Calmodulin 2 (P0DP24 Met1-Lys149). |
Conjugate | DyLight 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CALM CALM1 CALM3 CALML2 calmodulin calmodulin 2 (phosphorylase kinase delta) CAM CAM1 CAM2 CAM3 CAMB CAMC CAMII CAMIII FLJ99410 LP7057 protein PHKD PHKD2 phosphorylase kinase delta; calmodulin 2 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; LP7057 protein; phosphorylase kinase delta; phosphorylase kinase subunit delta; prepro-calmodulin 2
Gene Aliases: CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CAMB; CAMC; CAMII; CAMIII; LQT15; PHKD; PHKD2
UniProt ID: (Human) P0DP23
Entrez Gene ID: (Human) 805
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.