Novus Biologicals
Manufacturer Code:NB1001830FR
Catalog # NB1001830FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | HL 1499 |
Immunogen | N-terminal region of human Calcium Sensing Receptor. [UniProt# P41180] |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: calcium-sensing receptor; calcium-sensing receptor CAR CaSR EIG8 extracellular calcium-sensing receptor FHH FIH GPRC2AHHC HHC1 hypocalciuric hypercalcemia 1 NSHPT parathyroid Ca(2+)-sensing receptor 1 Parathyroid cell calcium-sensing receptor PCaR1 PCAR1MGC138441; CaR; Extracellular calcium-sensing receptor; parathyroid Ca(2+)-sensing receptor 1; Parathyroid cell calcium-sensing receptor 1; PCaR1
Gene Aliases: CAR; CASR; EIG8; FHH; FIH; GPRC2A; HHC; HHC1; HYPOC1; NSHPT; PCAR1
UniProt ID: (Human) P41180
Entrez Gene ID: (Human) 846
Molecular Function: G-protein coupled receptor ion channel ionotropic glutamate receptor ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.