R&D Systems
Manufacturer Code:MAB9608SP
Catalog # MAB9608SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 611825 |
Immunogen | Mouse myeloma cell line NS0-derived transfected with recombinant human Calcium-sensing R/CaSR protein Tyr20-Lys601 Accession # P41180-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: calcium-sensing receptor; calcium-sensing receptor CAR CaSR EIG8 extracellular calcium-sensing receptor FHH FIH GPRC2A GPRC2AHHC HHC1 hypocalciuric hypercalcemia 1 NSHPT parathyroid Ca(2&plus)-sensing receptor 1 Parathyroid cell calcium-sensing receptor PCaR1 PCAR1MGC138441; CaR; Extracellular calcium-sensing receptor; parathyroid Ca(2+)-sensing receptor 1; Parathyroid cell calcium-sensing receptor 1; PCaR1
Gene Aliases: CAR; CASR; EIG8; FHH; FIH; GPRC2A; HHC; HHC1; HYPOC1; NSHPT; PCAR1
UniProt ID: (Human) P41180
Entrez Gene ID: (Human) 846
Molecular Function:
G-protein coupled receptor
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.