Novus Biologicals
Manufacturer Code:NB12019347SS
Catalog # NB12019347T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5C10,ADD |
Immunogen | Synthetic peptide corresponding to residues (214) A D D D Y G R P G I E K F RE E A E E R D I (235) of human calcium sensing receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: calcium-sensing receptor; calcium-sensing receptor CAR CaSR EIG8 extracellular calcium-sensing receptor FHH FIH GPRC2AHHC HHC1 hypocalciuric hypercalcemia 1 NSHPT parathyroid Ca(2+)-sensing receptor 1 Parathyroid cell calcium-sensing receptor PCaR1 PCAR1MGC138441; CaR; Extracellular calcium-sensing receptor; parathyroid Ca(2+)-sensing receptor 1; Parathyroid cell calcium-sensing receptor 1; PCaR1
Gene Aliases: CAR; CASR; EIG8; FHH; FIH; GPRC2A; HHC; HHC1; HYPOC1; NSHPT; PCAR1
UniProt ID: (Human) P41180
Entrez Gene ID: (Human) 846
Molecular Function:
G-protein coupled receptor
ion channel
ionotropic glutamate receptor
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.