R&D Systems
Manufacturer Code:MAB6720SP
Catalog # MAB6720SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 861206 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Calcium Activated Nucleotidase 1/CANT1 Gly80-Ile401 Accession # Q8WVQ1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Apyrase homolog; Apyrase homolog calcium activated nucleotidase 1 DBQD EC 3.6.1.6 Putative MAPK-activating protein PM09 Putative NF-kappa-B-activating protein 107 SCAN1 SCAN-1Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase SHAPY SHAPYmicromelic dwarfism with vertebral and metaphyseal abnormalities and advancedcarpotarsal ossification soluble Ca-activated nucleotidase isozyme 1 soluble calcium-activated nucleotidase 1 soluble calcium-activated nucleotidase SCAN-1; Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase; micromelic dwarfism with vertebral and metaphyseal abnormalities and advanced carpotarsal ossification; Putative MAPK-activating protein PM09; Putative NF-kappa-B-activating protein 107; SCAN-1; soluble Ca-activated nucleotidase, isozyme 1; Soluble calcium-activated nucleotidase 1; soluble calcium-activated nucleotidase SCAN-1
Gene Aliases: CANT1; DBQD; DBQD1; SCAN-1; SCAN1; SHAPY
UniProt ID: (Human) Q8WVQ1
Entrez Gene ID: (Human) 124583
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.