Invitrogen
Manufacturer Code:720369
Catalog # PI720369
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to Human CALCR (aa 479-493) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles.,store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcitonin Receptor; CT-R
Gene Aliases: CALCR; CRT; CT-R; CTR; CTR1
UniProt ID: (Human) P30988
Entrez Gene ID: (Human) 799
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.