Novus Biologicals
Manufacturer Code:NB12011496
Catalog # NB12011496
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 14A2 |
Immunogen | Recombinant fragment (Human). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Alpha-type CGRP; ATP2B2plasma membrane calcium ATPase ATPase Ca++ transporting plasma membrane 4 DKFZp686G08106 DKFZp686M088 EC 3.6.3 EC 3.6.3.8 Matrix-remodeling-associated protein 1 matrix-remodelling associated 1 MXRA1 Plasma membrane calcium ATPase isoform 4 plasma membrane calcium pump Plasma membrane calcium pump isoform 4 plasma membrane calcium-transporting ATPase 4 PMCA4b PMCA4PMCA4x sarcolemmal calcium pump; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin-related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; CCP; CGRP-I; Katacalcin; PDN-21
Gene Aliases: CALC1; CALCA; CGRP; CGRP-I; CGRP1; CT; KC; PCT
UniProt ID: (Human) P06881
Entrez Gene ID: (Human) 796
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.