Invitrogen
Manufacturer Code:PA516464
Catalog # PIPA516464
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from human calcitonin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-type CGRP; alpha-type CGRP calcitonin calcitonin 1 calcitonin gene-related peptide I calcitonin/calcitonin-related polypeptide alpha katacalcin; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; calcitonin gene related peptide I; Calcitonin gene-related peptide; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin-related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; CCP; CGRP-I; Katacalcin; PDN-21; prepro-alpha-calcitonin gene-related peptide; procalcitonin
Gene Aliases: CA; Cal1; CAL6; CALC; CALC1; CALCA; Calcitonin; CCALCI; CGRP; CGRP-1; CGRP-I; CGRP1; CGRPI; CT; Ctn; KC; PCT; RATCAL6
UniProt ID: (Dog) P41547, (Horse) Q9N0T2, (Human) P06881, (Rat) P01256, (Mouse) Q99JA0
Entrez Gene ID: (Dog) 403946, (Horse) 100033906, (Human) 796, (Sheep) 100294596, (Rat) 24241, (Mouse) 12310
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.