Invitrogen
Manufacturer Code:MA523933
Catalog # PIMA523933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 212306 |
Immunogen | E. coli-derived recombit human Calcineurin B Met1-Val170 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: calcineurin B, type I (19kDa); Calcineurin subunit B type 1; CALNB1 CNB CNB1 calcineurin B type I (19kDa) calcineurin subunit B type 1 protein phosphatase 2B regulatory subunit 1 protein phosphatase 2B regulatory subunit B alpha protein phosphatase 3 (formerly 2B) regulatory subunit B (19kD) alpha isoform (calcineurin B type I; PP2B beta 1; protein phospatase 3, regulatory subunit B, alpha isoform (calcineurin B, type I); protein phospatase 3, regulatory subunit B, alpha isoform,type 1; Protein phosphatase 2B regulatory subunit 1; protein phosphatase 2B regulatory subunit B alpha; protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform (calcineurin B, type I); protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform (calcineurin B, type I); protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform; protein phosphatase 3 regulatory subunit B alpha; Protein phosphatase 3 regulatory subunit B alpha isoform 1; protein phosphatase 3, regulatory subunit B, alpha; protein phosphatase 3, regulatory subunit B, alpha isoform (calcineurin B, type I)
Gene Aliases: CALNB1; CaNB1; CNA2; CNB; CNB1; MCIP1; PPP3R1
UniProt ID: (Human) P63098, (Mouse) Q63810, (Rat) P63100
Entrez Gene ID: (Human) 5534, (Mouse) 19058, (Rat) 29748
Molecular Function:
calcium-binding protein
calmodulin
hydrolase
intracellular calcium-sensing protein
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.