Invitrogen
Manufacturer Code:PA583859
Catalog # PIPA583859
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PPP3R1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcineurin B, type I (19kDa); Calcineurin subunit B type 1; CALNB1 CNB CNB1; Protein phosphatase 2B regulatory subunit 1; protein phosphatase 2B regulatory subunit B alpha; protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform (calcineurin B, type I); protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform (calcineurin B, type I); protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform; Protein phosphatase 3 regulatory subunit B alpha isoform 1; protein phosphatase 3, regulatory subunit B, alpha
Gene Aliases: CALNB1; CNA2; CNB; CNB1; PPP3R1
UniProt ID: (Human) P63098
Entrez Gene ID: (Human) 5534
Molecular Function:
calcium-binding protein
calmodulin
hydrolase
intracellular calcium-sensing protein
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.