Invitrogen
Manufacturer Code:PA577819
Catalog # PIPA577819
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Human Calcineurin A peptide (AA 264-283) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha isoform formerly PPP2B Calcineurin A alpha Calcineurin A1 CalcineurinA Calmodulin dependent calcineurin A subunit alpha isoform Calmodulin-dependent calcineurin A subunit alpha isoform CALN CALNA CALNA 1 CAM PRP catalytic subunit CCN 1 Protein phosphatase 2B catalytic subunitProtein phosphatase 3 (formerly 2B) catalytic subunit alpha isoform Protein phosphatase 3 catalytic subunit alpha isoform PPP3CA Protein phosphatase 3 catalytic subunit alpha isozyme Serine/threonine protein phosphatase 2B catalytic subunit alpha isoform; calcineurin A alpha; calcineurin catalytic subunit; Calcineurin subunit A alpha; Calmodulin-dependent calcineurin A subunit alpha isoform; CAM-PRP catalytic subunit; CNA alpha; PP2B alpha 1; PP2BA alpha; protein phosphatase 2B, catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform; protein phosphatase 3 catalytic subunit alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isoform; protein phosphatase 3, catalytic subunit, alpha isozyme; protein phosphatase 3, catalytic subunit, gamma isozyme; protein phosphatase-2Ba; serine/threonine protein kinase; Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Gene Aliases: 2900074D19Rik; CALN; CALNA; CALNA1; CCN1; CN; CNA; CNA1; PPP2B; PPP3CA
UniProt ID: (Human) Q08209, (Mouse) P63328, (Rat) P63329
Entrez Gene ID: (Dog) 403486, (Rabbit) 100009423, (Human) 5530, (Mouse) 19055, (Rat) 24674
Molecular Function:
calcium-binding protein
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.