Invitrogen
Manufacturer Code:PA529209
Catalog # PIPA529209
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 264 of Human Calcineurin A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcineurin A alpha; Calmodulin-dependent calcineurin A subunit alpha isoform; CALN CALNA CALNA1 CCN1 CNA1 PPP2B CAM-PRP catalytic subunit calcineurin A alpha calmodulin-dependent calcineurin A subunit alpha isoform protein phosphatase 2B catalytic subunit alpha isoform protein phosphatase 3 (formerly 2B) catalytic subunit alpha isoform serine/threonine-protein; CAM-PRP catalytic subunit; CNA alpha; PP2B alpha 1; PP2BA alpha; protein phosphatase 2B, catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform; protein phosphatase 3 catalytic subunit alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isozyme; Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Gene Aliases: 2900074D19Rik; CALN; CALNA; CALNA1; CCN1; CN; CNA; CNA1; PPP2B; PPP3CA
UniProt ID: (Human) Q08209, (Mouse) P63328
Entrez Gene ID: (Human) 5530, (Mouse) 19055
Molecular Function: calcium-binding protein hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.