Novus Biologicals
Manufacturer Code:NB11040552
Catalog # NB11040552
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Protein Phosphatase 3 Catalytic Subunit Alpha isoform using the numbering given in entry NP_000935.1 (GeneID 5530). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcineurin A alpha; calcineurin A alpha Calmodulin-dependent calcineurin A subunit alpha isoform CALN CALNAcatalytic subunit alpha isoform CAM-PRP catalytic subunit CNA CNA1CALNA1 EC 3.1.3.16 PPP2BCCN1 protein phosphatase 2B catalytic subunit alpha isoform protein phosphatase 3 (formerly 2B) catalytic subunit alpha isoform(calcineurin A alpha) protein phosphatase 3 catalytic subunit alpha isozyme serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform; Calmodulin-dependent calcineurin A subunit alpha isoform; CAM-PRP catalytic subunit; protein phosphatase 2B, catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform; protein phosphatase 3 catalytic subunit alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isozyme; Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Gene Aliases: CALN; CALNA; CALNA1; CCN1; CNA; CNA1; PPP2B; PPP3CA
UniProt ID: (Human) Q08209
Entrez Gene ID: (Human) 5530
Molecular Function: calcium-binding protein hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.