Abnova
Manufacturer Code:MAB1510
Catalog # 89-110-464
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CB-955 |
Immunogen | Native purified bovine calbindin-D. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C on dry atmosphere. After reconstitution with 1mL of 1.2% sodium acetate or neutral PBS to a concentration of 100&mug/mL,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against native calbindin-D.
For Research Use Only
Protein Aliases: Calbindin; calbindin 1, 28kDa; Calbindin D28; D-28K; MGC138959; Vitamin D-dependent calcium-binding protein, avian-type
Gene Aliases: CALB1
UniProt ID: (Bovine) P04467
Entrez Gene ID: (Bovine) 539930
Molecular Function:
calcium-binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.