Proteintech
Manufacturer Code:510591AP150UL
Catalog # 50-173-5877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Cadm1 Fusion Protein Ag0449 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900073G06Rik 3100001I08Rik AI987920 Bl2 Cadm1 cell adhesion molecule 1 Igsf4 Igsf4a Necl2 RA175A RA175B RA175C RA175N SgIGSF ST17 SynCam Tslc1; cadm1; Cell adhesion molecule 1; cell adhesion molecule 1 transcript variant 8abc; IgSF4; Immunoglobulin superfamily member 4; immunoglobulin superfamily member 4A; immunosuperfamily protein Bl2; NECL-2; nectin-like 2; Nectin-like protein 2; SgIgSF; Spermatogenic immunoglobulin superfamily; Synaptic cell adhesion molecule; SynCAM; TSLC-1; Tumor suppressor in lung cancer 1
Gene Aliases: 2900073G06Rik; 3100001I08Rik; AI987920; Bl2; Cadm1; Igsf4; Igsf4a; Necl2; RA175; RA175A; RA175B; RA175C; RA175N; SgIGSF; ST17; SynCam; SynCam1; Tslc1
UniProt ID: (Mouse) Q8R5M8
Entrez Gene ID: (Mouse) 54725
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.