R&D Systems
Manufacturer Code:AF3264SP
Catalog # AF3264SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Cadherin-13 Glu23-Ala692 Accession # P55290 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cadherin 13 H-cadherin (heart) cadherin-13 Cadherin13 CDH13 CDHHT-cadherin H-cadherin Heart cadherin heart-cadherin P105 T-cad T-Cadherin Truncated cadherin truncated-cadherin; cadherin 13, H-cadherin (heart); Cadherin-13; H-cadherin; H-cadherin (heart); Heart cadherin; P105; T-cad; T-cadherin; Truncated cadherin
Gene Aliases: CDH13; CDHH; P105
UniProt ID: (Human) P55290
Entrez Gene ID: (Human) 1012
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.