R&D Systems
Manufacturer Code:FAB17901G
Catalog # FAB17901G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 667039 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Cadherin-11 Phe23-Thr617 Accession # AAA35622 |
Conjugate | Alexa Fluor 488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CAD11OSF-4 cadherin 11 type 2 OB-cadherin (osteoblast) cadherin-11 Cadherin11 CDH11 CDHOB OB OB-cadherin Osteoblast cadherin; cadherin 11, type 2, OB-cadherin (osteoblast); Cadherin-11; OB-cadherin; OSF-4; Osteoblast cadherin
Gene Aliases: CAD11; CDH11; CDHOB; OB; OSF-4
UniProt ID: (Human) P55287
Entrez Gene ID: (Human) 1009
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.