R&D Systems
Manufacturer Code:MAB1790
Catalog # MAB1790
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 283416 |
Immunogen | <EM>S. frugiperda</EM> insect ovarian cell line <EM>Sf</EM> 21-derived recombinant human Cadherin-11 Phe23-Thr617 Accession # AAA35622 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CAD11OSF-4 cadherin 11 type 2 OB-cadherin (osteoblast) cadherin-11 Cadherin11 CDH11 CDHOB OB OB-cadherin Osteoblast cadherin; cadherin 11, type 2, OB-cadherin (osteoblast); Cadherin-11; OB-cadherin; OSF-4; Osteoblast cadherin
Gene Aliases: CAD11; CDH11; CDHOB; OB; OSF-4
UniProt ID: (Human) P55287
Entrez Gene ID: (Human) 1009
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.