Abnova
Manufacturer Code:MAB7895
Catalog # 89-113-216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S8b-1 |
Immunogen | A synthetic peptide corresponding to amino acids 189-205 of rat Kcnt1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of Kcnt1.
For Research Use Only
Protein Aliases: CAB2; Cacnlb2; Calcium channel voltage-dependent subunit beta 2; Voltage-dependent L-type calcium channel subunit beta-2
Gene Aliases: Cacnb2; Cacnlb2
UniProt ID: (Rat) Q8VGC3
Entrez Gene ID: (Rat) 116600
Molecular Function: calcium channel ion channel transporter voltage-gated calcium channel voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.