Abnova
Manufacturer Code:PAB9800
Catalog # 89-134-427
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to rat Cacnb2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Cacnb2.
For Research Use Only
Protein Aliases: CAB2; Cacnlb2; Calcium channel voltage-dependent subunit beta 2; Voltage-dependent L-type calcium channel subunit beta-2
Gene Aliases: Cacnb2; Cacnlb2
UniProt ID: (Rat) Q8VGC3
Entrez Gene ID: (Rat) 116600
Molecular Function:
calcium channel
ion channel
transporter
voltage-gated calcium channel
voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.