Abnova
Manufacturer Code:MAB11552
Catalog # 89-109-703
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S178A-9 |
Immunogen | Recombinant fusion protien corresponding to amino acids 2052-2172 at C-terminus of mouse Cacna1g. |
Conjugate | PE/ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant of Cacna1g.
For Research Use Only
Protein Aliases: [a]1G; a1G; alpha-1G; Cav3.1d; mKIAA1123; voltage-dependent T-type calcium channel subunit alpha-1G
Gene Aliases: [a]1G; a1G; alpha-1G; Cav3.1d; mKIAA1123
Entrez Gene ID: (Mouse) 12291
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.