Invitrogen
Manufacturer Code:PA599755
Catalog # PIPA599755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CACNA1G corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [a]1G a1G alpha-1G Ca(V)T.1 CACNA1G calcium channel voltage-dependent T type alpha 1G subunit calcium voltage-gated channel subunit alpha1 G CaV T1 Cav3 1 cav3 1c Cav3.1 cav3.1c Cav3.1d KIAA1123 MGC117234 mKIAA1123 NBR13 SCA42 voltage-dependent calcium channel alpha 1G subunit isoform 11 voltage-dependent T-type calcium channel alpha 1G subunit voltage-dependent T-type calcium channel subunit alpha-1G Voltage-gated calcium channel subunit alpha Cav3.1; calcium channel, voltage-dependent, T type, alpha 1G subunit; Cav3.1c; NBR13; voltage-dependent calcium channel alpha 1G subunit, isoform 11; voltage-dependent T-type calcium channel alpha 1G subunit; Voltage-dependent T-type calcium channel subunit alpha-1G; Voltage-gated calcium channel subunit alpha Cav3.1
Gene Aliases: [a]1G; a1G; alpha-1G; Ca(V)T.1; CACNA1G; Cav3.1; Cav3.1d; KIAA1123; mKIAA1123; NBR13; SCA42
UniProt ID: (Human) Q19R12
Entrez Gene ID: (Human) 8913, (Rat) 29717, (Mouse) 12291
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.