Invitrogen
Manufacturer Code:PA575644
Catalog # PIPA575644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human Calcium Channel 1E. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BII; Brain calcium channel II; Calcium channel, L type, alpha-1 polypeptide, isoform 6; calcium channel, R type, alpha-1 polypeptide, isoform 6; calcium channel, voltage-dependent, L type, alpha 1E subunit; RBE-II; RBE2; voltage gated calcium channel alpha1E subunit; voltage-dependent calcium channel alpha 1E subunit; voltage-dependent calcium channel alpha1-E subunit; Voltage-dependent R-type calcium channel subunit alpha-1E; Voltage-dependent R-type calcium channel subunit alpha-1E Brain calcium channel II BII Calcium channel L type alpha-1 polypeptide isoform 6 Voltage-gated calcium channel subunit alpha Cav2.3 CACNA1E CACH6 CACNL1A6; voltage-gated calcium channel alpha subunit Cav2.3; Voltage-gated calcium channel subunit alpha Cav2.3
Gene Aliases: A430040I15; alpha1E; BII; CACH6; CACHA1E; CACNA1E; CACNL1A6; Cav2.3; Cchra1; gm139
UniProt ID: (Human) Q15878
Entrez Gene ID: (Human) 777, (Rat) 54234, (Mouse) 12290
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.