Invitrogen
Manufacturer Code:PA577361
Catalog # PIPA577361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)EQLTKETEGGNHS corresponding to amino acid residues 215-227 of rat CaV 1.3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha1D; brain class D; calcium channel alpha-1 subunit; Calcium channel, L type, alpha-1 polypeptide isoform 2; Calcium channel, L type, alpha-1 polypeptide, isoform 2; calcium channel, neuroendocrine/brain-type, alpha 1 subunit; calcium channel, voltage-dependent, L type, alpha 1D subunit; CaV1.3alpha1; Rat brain class D; RBD; voltage-dependent calcium channel subunit alpha1D; Voltage-dependent L-type calcium channel subunit alpha-1D; voltage-gated calcium channel alpha 1 subunit; voltage-gated calcium channel alpha subunit Cav1.3; voltage-gated calcium channel pore forming subunit CaV1.3alpha1 IVS3-IVS4 extracellular linker; Voltage-gated calcium channel subunit alpha Cav1.3
Gene Aliases: 8430418G19Rik; C79217; CACH3; CACN4; CACNA1D; CACNL1A2; Cav1.3; Cchl1a; CCHL1A2; D-LTCC; PASNA; SANDD
UniProt ID: (Human) Q01668, (Mouse) Q99246, (Rat) P27732
Entrez Gene ID: (Human) 776, (Mouse) 12289, (Rat) 29716
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.