Novus Biologicals
Manufacturer Code:NBP237428
Catalog # NBP237428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8C5B8 |
Immunogen | Purified recombinant fragment of human CAMK4 (AA: 35-292 ) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: brain Ca(2+)-calmodulin-dependent protein kinase type IV; brain Ca(2+)-calmodulin-dependent protein kinase type IV brain Ca++-calmodulin-dependent protein kinase type IV calcium/calmodulin-dependent protein kinase IV calcium/calmodulin-dependent protein kinase type IV calcium/calmodulin-dependent protein kinase type IV catalytic chain CAM kinase- GR CAM kinase IV CaM kinase-GR caMK CaMK IV CaMK-GR EC 2.7.11 EC 2.7.11.17 IV MGC36771; brain Ca++-calmodulin-dependent protein kinase type IV; calcium/calmodulin-dependent protein kinase IV; Calcium/calmodulin-dependent protein kinase type IV; calcium/calmodulin-dependent protein kinase type IV catalytic chain; CAM kinase IV; CAM kinase- GR; CaM kinase-GR; CaMK IV
Gene Aliases: CAMK; CaMK IV; CAMK-GR; CAMK4; CAMKIV
UniProt ID: (Human) Q16566
Entrez Gene ID: (Human) 814
Molecular Function:
cytoskeletal protein
kinase
microtubule family cytoskeletal protein
non-motor microtubule binding protein
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.