Invitrogen
Manufacturer Code:MA530168
Catalog # PIMA530168
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 162 |
Immunogen | Recombit Mouse CaM Kinase 4/CaMKIV protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: Ca2+/calmodulin-dependent protein kinase type IV/Gr; calcium/calmodulin-dependent protein kinase type IV; CAM kinase-GR; caMK IV; CaMKIV Brain Ca(2+) calmodulin dependent protein kinase type Calcium / calmodulin dependent protein kinase type 4 catalytic chain Calcium/calmoduli
Gene Aliases: A430110E23Rik; AI666733; CaMKIV; CaMKIV/Gr; D18Bwg0362e
Entrez Gene ID: (Mouse) 12326
Molecular Function:
cytoskeletal protein
kinase
microtubule family cytoskeletal protein
non-motor microtubule binding protein
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.