Invitrogen
Manufacturer Code:PA517690
Catalog # PIPA517690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide around amino acid 282-300 of human CaMKIV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: brain Ca(2+)-calmodulin-dependent protein kinase type IV; brain Ca++-calmodulin-dependent protein kinase type IV; Ca2+/calmodulin-dependent protein kinase type IV/Gr; calcium/calmodulin-dependent protein kinase IV; Calcium/calmodulin-dependent protein kinase type IV; calcium/calmodulin-dependent protein kinase type IV catalytic chain; Calmodulin-dependent protein kinase IV; Calspermin; CAM kinase IV; CAM kinase IV CAM kinase- GR OTTHUMP00000222818 brain Ca(2+)-calmodulin-dependent protein kinase type IV brain Ca++-calmodulin-dependent protein kinase type IV caM kinase-GR CaMK IV calcium/calmodulin-dependent protein kinase type IV calcium/calmodulin-dependent protein kinase type IV catalytic chain; CAM kinase- GR; CAM kinase-GR; CaMK IV
Gene Aliases: A430110E23Rik; AI666733; CAMK; CaMK IV; CAMK-GR; CAMK4; CAMKIV; CaMKIV/Gr; Ccdpk; D18Bwg0362e; RATCCDPK
UniProt ID: (Human) Q16566, (Rat) P13234
Entrez Gene ID: (Human) 814, (Mouse) 12326, (Rat) 25050
Molecular Function:
cytoskeletal protein
kinase
microtubule family cytoskeletal protein
non-motor microtubule binding protein
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.