Invitrogen
Manufacturer Code:PA565276
Catalog # PIPA565276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human CaMKII gamma |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II gamma; Calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II subunit gamma; caMK-II subunit gamma; CAMK2B CAMK2D CAMK2G Calcium/calmodulin-dependent protein kinase type II subunit beta CaM kinase II subunit beta CaMK-II subunit beta CAMK2B CAM2 CAMK2 CAMKB Calcium/calmodulin-dependent protein kinase type II subunit gamma CaM kinase II subunit gamma CaMK-II subunit gamma CAMK2G CAMKG Calcium/calmodulin-dependent protein kinase type II subunitdelta CaM kinase II subunit delta CaMK-II subunit delta CAMK2D CAMKD
Gene Aliases: CAMK; CAMK-II; CAMK2G; CAMKG
UniProt ID: (Human) Q13555
Entrez Gene ID: (Human) 818
Molecular Function: non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.