Proteintech
Manufacturer Code:126662AP150UL
Catalog # 50-172-8157
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CaMKII gamma Fusion Protein Ag3348 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ca2+/calmodulin-dependent protein kinase II; calcium/calmodulin-dependent protein kinase (CaM kinase) II gamma; calcium/calmodulin-dependent protein kinase II gamma; calcium/calmodulin-dependent protein kinase type II gamma chain; Calcium/calmodulin-dependent protein kinase type II subunit gamma; CaM kinase II subunit gamma; CaM kinase II subunit gamma CAMK CAMK II CaMK II subunit gamma CAMK2G CAMKG; caM-kinase II gamma chain; CaMK II; caMK-II subunit gamma
Gene Aliases: CAMK; CAMK-II; CAMK2G; CAMKG
UniProt ID: (Human) Q13555, (Mouse) Q923T9, (Rat) P11730
Entrez Gene ID: (Human) 818, (Mouse) 12325, (Rat) 171140
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.